Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Dimethyl (trityloxyimino)malonate, 98%
Click to view available options
Quantity:
250mg
1g
5g
Specifications
Quantity | 250mg |
Formula Weight | 403.43 |
Physical Form | Solid |
Percent Purity | 98% |
Chemical Name or Material | Dimethyl (trityloxyimino)malonate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures