Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylamine, 2M solution in methyl alcohol, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
124-40-3,67-56-1
C2H7N
45.085
ROSDSFDQCJNGOL-UHFFFAOYSA-N
dimethylamine,n,n-dimethylamine,methanamine, n-methyl,dimethylamin,dimethyl amine,dimethyl-amine,dimethylamine anhydrous,rcra waste number u092,dimethylamine solution
674
CHEBI:17170
N-methylmethanamine
CNC
124-40-3,67-56-1
C2H7N
45.085
ROSDSFDQCJNGOL-UHFFFAOYSA-N
dimethylamine,n,n-dimethylamine,methanamine, n-methyl,dimethylamin,dimethyl amine,dimethyl-amine,dimethylamine anhydrous,rcra waste number u092,dimethylamine solution
674
CHEBI:17170
N-methylmethanamine
CNC
Quantity 800mL
Packaging AcroSeal™ Glass bottle
Formula Weight 45.08
Chemical Name or Material Dimethylamine, 2M solution in methanol, Acros Organics™