Learn More
Dimethylamine, 2M solution in methyl alcohol, AcroSeal™, Thermo Scientific Chemicals
$270.00
Specifications
CAS | 124-40-3, 67-56-1 |
---|---|
Packaging | AcroSeal™ Glass bottle |
Synonym | dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution |
InChI Key | ROSDSFDQCJNGOL-UHFFFAOYSA-N |
SMILES | CNC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC370261000
|
Thermo Scientific Chemicals
370261000 |
100 mL | AcroSeal™ Glass bottle |
Each for $86.00
Order in Bulk
Request bulk or custom formats |
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
AC370268000
|
Thermo Scientific Chemicals
370268000 |
800 mL | AcroSeal™ Glass bottle |
Each for $270.00
Order in Bulk
Request bulk or custom formats |
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
124-40-3, 67-56-1 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
CNC | |
674 | |
45.08 |
AcroSeal™ Glass bottle | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
CHEBI:17170 |
Safety and Handling
GHS H Statement
Causes damage to organs if inhaled.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable l
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 204-697-4
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.