Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Dimethylamino-3',4'-dihydroxyacetophenone, Hydrochloride 98%, Acros Organics

Click to view available options
Quantity:
500mg
16899-83-5
C10H14ClNO3
231.676
MFCD00044781
LVRPQCAWIVYJTP-UHFFFAOYSA-N
1-3,4-dihydroxyphenyl-2-dimethylamino ethanone hydrochloride,1-3,4-dihydroxyphenyl-2-dimethylamino ethan-1-one hydrochloride,2-dimethylamino-3',4'-dihydroxyacetophenone hydrochloride,alpha-dimethylamino-3,4'-dihydroxyacetophenone hydrochloride,alpha-dimetgylamino-3',4'-dihydroxyacetophenone hydrochloride,ethanone,4-dihydroxyphenyl-2-dimethylamino-, hydrochloride
54599386
1-(3,4-dihydroxyphenyl)-2-(dimethylamino)ethanone;hydrochloride
CN(C)CC(=O)C1=CC(=C(C=C1)O)O.Cl
16899-83-5
C10H14ClNO3
231.676
MFCD00044781
LVRPQCAWIVYJTP-UHFFFAOYSA-N
1-3,4-dihydroxyphenyl-2-dimethylamino ethanone hydrochloride,1-3,4-dihydroxyphenyl-2-dimethylamino ethan-1-one hydrochloride,2-dimethylamino-3',4'-dihydroxyacetophenone hydrochloride,alpha-dimethylamino-3,4'-dihydroxyacetophenone hydrochloride,alpha-dimetgylamino-3',4'-dihydroxyacetophenone hydrochloride,ethanone,4-dihydroxyphenyl-2-dimethylamino-, hydrochloride
54599386
1-(3,4-dihydroxyphenyl)-2-(dimethylamino)ethanone;hydrochloride
CN(C)CC(=O)C1=CC(=C(C=C1)O)O.Cl
Infrared Spectrum Authentic
Color Gray
Quantity 500mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 231.22
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material α-Dimethylamino-3', 4'-dihydroxyacetophenone, hydrochloride, 98%