Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylaminoacetaldehyde dimethylacetal, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
38711-20-5
C6H15NO2
133.19
MFCD00010205
HUYAEQCJNXODLQ-UHFFFAOYSA-N
ethanamine, 2,2-dimethoxy-n,n-dimethyl,dimethylamino acetaldehyde dimethyl acetal,dimethylaminoacetaldehyde dimethyl acetal,dimethylaminoacetaldehyde dimethylacetal,n-2,2-dimethoxyethyl dimethylamine,2-dimethylamino acetaldehyde dimethylacetal,2,2-dimethoxyethyl dimethylamine,2,2-dimethoxyethyl dimethyl amine,2,2-dimethoxy-n,n-dimethylethanamin,acmc-209j1l
123476
2,2-dimethoxy-N,N-dimethylethanamine
CN(C)CC(OC)OC
38711-20-5
C6H15NO2
133.19
MFCD00010205
HUYAEQCJNXODLQ-UHFFFAOYSA-N
ethanamine, 2,2-dimethoxy-n,n-dimethyl,dimethylamino acetaldehyde dimethyl acetal,dimethylaminoacetaldehyde dimethyl acetal,dimethylaminoacetaldehyde dimethylacetal,n-2,2-dimethoxyethyl dimethylamine,2-dimethylamino acetaldehyde dimethylacetal,2,2-dimethoxyethyl dimethylamine,2,2-dimethoxyethyl dimethyl amine,2,2-dimethoxy-n,n-dimethylethanamin,acmc-209j1l
123476
2,2-dimethoxy-N,N-dimethylethanamine
CN(C)CC(OC)OC
Boiling Point 64°C to 66°C (12.0mmHg)
Specific Gravity 0.9
Flash Point 32°C
Density 0.9000g/mL
Refractive Index 1.4100 to 1.412
Color Colorless to Yellow
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (CH3)2NCH2CH(OCH3)2
Formula Weight 133.19
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Dimethylaminoacetaldehyde dimethylacetal
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger