Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Dimethylaminoacetonitrile, 98%
Click to view available options
Quantity:
100g
Specifications
Density | 0.863 |
Refractive Index | 1.41 |
Boiling Point | 137°C to 138°C |
Quantity | 100g |
Flash Point | 36°C (96°F) |
Odor | Recognizable |
Linear Formula | (CH3)2NCH2CN |
UN Number | UN2378 |
Beilstein | 1735677 |
Formula Weight | 84.12 |
Show More |
Safety and Handling
GHS H Statement
H300-H310-H226-H319
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P262-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P363-P370+P378q-P501c
H226-H301+H331-H310-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOACETONITRILE
EINECSNumber : 213-140-4
RTECSNumber : AL9450000
TSCA : No
Recommended Storage : Ambient temperatures