Alfa Aesar™ Dimethylaminoacetonitrile, 98%

Manufacturer: Alfa Aesar™ B2508514

Quantity 25g

 View more versions of this product

Catalog No. AAB2508514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 926-64-7
    Molecular Formula C4H8N2
    Linear Formula (CH3)2NCH2CN
    Formula Weight 84.12
    MDL Number MFCD00001890
    UN Number UN2378
    Beilstein 1735677
    Chemical Name or Material Dimethylaminoacetonitrile

    Specifications

    Formula Weight 84.12
    Flash Point 36°C (96°F)
    Molecular Formula C4H8N2
    Linear Formula (CH3)2NCH2CN
    Boiling Point 137° to 138°C
    MDL Number MFCD00001890
    CAS 926-64-7
    UN Number UN2378
    Beilstein 1735677
    Quantity 25g
    Odor Recognizable
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H310-H226-H319
    Fatal if swallowed.
    Fatal in contact with skin.
    Flammable liquid and vapour.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310-P302+P352
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOACETONITRILE
    RTECS Number AL9450000
    Chemical Name or Material Dimethylaminoacetonitrile
    Assay 98%

    GHS H Statement
    H300-H310-H226-H319
    Fatal if swallowed.
    Fatal in contact with skin.
    Flammable liquid and vapour.
    Causes serious eye irritation.

    GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310-P302+P352
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit