Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylanilinium Tetrakis (pentafluorophenyl)borate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
118612-00-3
C32H12BF20N
801.23
MFCD01074420
BRHZQNMGSKUUMN-UHFFFAOYSA-O
dimethylanilinium tetrakis pentafluorophenyl borate,unii-h8r86l92nd,n,n-dimethylanilinium tetrakis pentafluorophenyl borate,n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate,n,n-dimethylanilinium tetra pentafluorophenyl borate,benzene, pentafluoro-, boron complex,n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-,benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-,dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate,borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1
10996402
dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1
118612-00-3
C32H12BF20N
801.23
MFCD01074420
BRHZQNMGSKUUMN-UHFFFAOYSA-O
dimethylanilinium tetrakis pentafluorophenyl borate,unii-h8r86l92nd,n,n-dimethylanilinium tetrakis pentafluorophenyl borate,n,n-dimethylbenzenaminium tetrakis perfluorophenyl borate,n,n-dimethylanilinium tetra pentafluorophenyl borate,benzene, pentafluoro-, boron complex,n,n-dimethylanilinium tetrakis pentafluorophenyl borate 1-,benzenamine, n,n-dimethyl-, tetrakis pentafluorophenyl borate 1-,dimethyl phenyl ammonium tetrakis 2,3,4,5,6-pentafluorophenyl borate,borate 1-, tetrakis pentafluorophenyl-, hydrogen, compd. with n,n-dimethylbenzenamine 1:1:1
10996402
dimethyl(phenyl)azanium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.C[NH+](C)C1=CC=CC=C1
Quantity 1g
NMR 97.5% min.
Packaging Glass bottle
Color Blue-White
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 801.23
Physical Form Powder, s On Exposure To Air
Percent Purity 98%
Chemical Name or Material Dimethylanilinium tetrakis (pentafluorophenyl)borate

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
Suspected of causing cancer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical

GHS Signal Word: Danger