Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylcyanamide, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
Packaging:
Glass bottle
1467-79-4
C3H6N2
70.09
MFCD00001767
OAGOUCJGXNLJNL-UHFFFAOYSA-N
dimethyl cyanamide,cyanamide, dimethyl,n-cyano-n-methylmethanamine,dimethylkyanamid,cyanodimethylamine,n-cyanodimethylamine,dimethylkyanamid czech,unii-6p25ipq8gn,ccris 5909,cyanamide, n,n-dimethyl
15112
dimethylcyanamide
CN(C)C#N
1467-79-4
C3H6N2
70.09
MFCD00001767
OAGOUCJGXNLJNL-UHFFFAOYSA-N
dimethyl cyanamide,cyanamide, dimethyl,n-cyano-n-methylmethanamine,dimethylkyanamid,cyanodimethylamine,n-cyanodimethylamine,dimethylkyanamid czech,unii-6p25ipq8gn,ccris 5909,cyanamide, n,n-dimethyl
15112
dimethylcyanamide
CN(C)C#N
Specific Gravity 0.867
Density 0.8670g/mL
Boiling Point 161.0°C to 163.0°C
Color Colorless to Yellow
Melting Point -41.0°C
Packaging Glass bottle
Quantity 100mL
Infrared Spectrum Authentic
Refractive Index 1.4090 to 1.4110
Water 0.1% max. (K.F.)
Flash Point 58°C
Assay Percent Range 97%
Linear Formula (CH3)2NCN
Beilstein 04, 74
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone,ethyl ether,ethnaol
Formula Weight 70.09
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dimethylcyanamide, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f

GHS Signal Word: Danger

EINECSNumber : 215-991-7

RTECSNumber : GS6475000

TSCA : TSCA