Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyldi-n-octadecylammonium chloride, 96%

Manufacturer:  Alfa Aesar™ B2378818

Quantity 50g

 View more versions of this product

Catalog No. AAB2378818

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

107-64-2
[CH3(CH2)17]2N(CH3)2Cl
MFCD00055540
3580288
Dimethyldi-n-octadecylammonium chloride
C38H80ClN
586.51
UN3077
Dimethyldistearylammonium chloride

Specifications

107-64-2
[CH3(CH2)17]2N(CH3)2Cl
MFCD00055540
3580288
Odorless
Dimethyldi-n-octadecylammonium chloride
C38H80ClN
586.51
UN3077
50g
Dimethyldistearylammonium chloride
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : BQ1923000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit