Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl (S)-(-)-malate, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
617-55-0
C6H10O5
162.14
YSEKNCXYRGKTBJ-BYPYZUCNSA-N
s-dimethyl 2-hydroxysuccinate,dimethyl s---malate,dimethyl l-malate,l---malic acid dimethyl ester,1,4-dimethyl 2s-2-hydroxybutanedioate,butanedioic acid, hydroxy-, dimethyl ester, 2s,dimethyl l---malate,pubchem6778,di-methyl s-malate,dimethyl s--malate
10285815
dimethyl (2S)-2-hydroxybutanedioate
COC(=O)CC(C(=O)OC)O
617-55-0
C6H10O5
162.14
YSEKNCXYRGKTBJ-BYPYZUCNSA-N
s-dimethyl 2-hydroxysuccinate,dimethyl s---malate,dimethyl l-malate,l---malic acid dimethyl ester,1,4-dimethyl 2s-2-hydroxybutanedioate,butanedioic acid, hydroxy-, dimethyl ester, 2s,dimethyl l---malate,pubchem6778,di-methyl s-malate,dimethyl s--malate
10285815
dimethyl (2S)-2-hydroxybutanedioate
COC(=O)CC(C(=O)OC)O
Specific Gravity 1.23
Density 1.2300g/mL
Refractive Index 1.4366
Packaging Glass bottle
Specific Rotation − 6.70
Flash Point 113°C
Quantity 5g
Boiling Point 91.0°C to 93.0°C (1.0 Torr)
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula H3COOCCH(OH)CH2COOCH3
Solubility Information Solubility in water: in water:(20 °C) insoluble.
Formula Weight 162.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Dimethyl (S)-(-)-malate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning