Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 1,3-acetonedicarboxylate, 97%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1830-54-2
C7H10O5
174.15
RNJOKCPFLQMDEC-UHFFFAOYSA-N
dimethyl 1,3-acetonedicarboxylate,dimethyl acetone-1,3-dicarboxylate,dimethyl 3-oxoglutarate,1,5-dimethyl 3-oxopentanedioate,pentanedioic acid, 3-oxo-, dimethyl ester,dimethyl acetonedicarboxylate,acetone dicarboxylic acid, dimethyl ester,glutaric acid, 3-oxo-, dimethyl ester,pentanedioic acid, 3-oxo-, 1,5-dimethyl ester,dimethyl 3-oxopentane-1,5-dioate
74591
dimethyl 3-oxopentanedioate
COC(=O)CC(=O)CC(=O)OC
1830-54-2
C7H10O5
174.15
RNJOKCPFLQMDEC-UHFFFAOYSA-N
dimethyl 1,3-acetonedicarboxylate,dimethyl acetone-1,3-dicarboxylate,dimethyl 3-oxoglutarate,1,5-dimethyl 3-oxopentanedioate,pentanedioic acid, 3-oxo-, dimethyl ester,dimethyl acetonedicarboxylate,acetone dicarboxylic acid, dimethyl ester,glutaric acid, 3-oxo-, dimethyl ester,pentanedioic acid, 3-oxo-, 1,5-dimethyl ester,dimethyl 3-oxopentane-1,5-dioate
74591
dimethyl 3-oxopentanedioate
COC(=O)CC(=O)CC(=O)OC
Flash Point >110°C
Specific Gravity 1.18
Packaging Glass bottle
Melting Point 16.0°C to 17.0°C
Quantity 25g
Boiling Point 237.0°C to 241.0°C
Infrared Spectrum Authentic
Color Colorless to Yellow
Refractive Index 1.4430 to 1.4450
Density 1.1800g/mL
Assay Percent Range 97%
Linear Formula CH3O2CCH2COCH2CO2CH3
Beilstein 03,79
Solubility Information Solubility in water: 120g/L (20°C). Other solubilities: immiscible with ligroin and cyclohexane,soluble in chloroform and ethyl acetate,soluble in acetone,methanol,143g/L 1 n naoh,165g/L 10 % acetic acid
Formula Weight 174.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dimethyl 1, 3-acetonedicarboxylate
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning