Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl acetylenedicarboxylate, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
762-42-5
C6H6O4
142.11
VHILMKFSCRWWIJ-UHFFFAOYSA-N
dimethyl acetylenedicarboxylate,2-butynedioic acid, dimethyl ester,1,4-dimethyl but-2-ynedioate,dimethyl butynedioate,dimethyl 2-butynedioate,acetylenedicarboxylic acid dimethyl ester,dmad,methyl acetylenedicarboxylate,bis methoxycarbonyl acetylene,di carbomethoxy acetylene
12980
dimethyl but-2-ynedioate
COC(=O)C#CC(=O)OC
762-42-5
C6H6O4
142.11
VHILMKFSCRWWIJ-UHFFFAOYSA-N
dimethyl acetylenedicarboxylate,2-butynedioic acid, dimethyl ester,1,4-dimethyl but-2-ynedioate,dimethyl butynedioate,dimethyl 2-butynedioate,acetylenedicarboxylic acid dimethyl ester,dmad,methyl acetylenedicarboxylate,bis methoxycarbonyl acetylene,di carbomethoxy acetylene
12980
dimethyl but-2-ynedioate
COC(=O)C#CC(=O)OC
Alkylation reagent
Specific Gravity 1.16
Boiling Point 95.0°C to 98.0°C (19.0mmHg)
Flash Point 86°C
Quantity 500g
Infrared Spectrum Authentic
Melting Point -18.0°C
Refractive Index 1.4460 to 1.4480
Density 1.1600g/mL
Color Colorless to Yellow
Assay Percent Range 98%
Linear Formula CH3O2CC≡CCO2CH3
Fieser 01,272; 02,145; 03,103; 04,168; 05,227; 06,206; 07,117; 09,176
Formula Weight 142.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Dimethyl acetylenedicarboxylate
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger