Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl carbonate, 99%, Acros Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
616-38-6
C3H6O3
90.08
MFCD00008420
IEJIGPNLZYLLBP-UHFFFAOYSA-N
methyl carbonate,carbonic acid, dimethyl ester,carbonic acid dimethyl ester,methyl carbonate meo 2co,unii-ke9j097spn,dimethylcarbonate,ch3ocooch3,ke9j097spn,dimethyl ester of carbonic acid,dsstox_cid_9192
12021
CHEBI:36596
dimethyl carbonate
COC(=O)OC
616-38-6
C3H6O3
90.08
MFCD00008420
IEJIGPNLZYLLBP-UHFFFAOYSA-N
methyl carbonate,carbonic acid, dimethyl ester,carbonic acid dimethyl ester,methyl carbonate meo 2co,unii-ke9j097spn,dimethylcarbonate,ch3ocooch3,ke9j097spn,dimethyl ester of carbonic acid,dsstox_cid_9192
12021
CHEBI:36596
dimethyl carbonate
COC(=O)OC
Alkylation reagent
Specific Gravity 1.069
Viscosity 0.625 mPa.s (20°C)
Packaging Plastic drum
Flash Point 18°C
Density 1.0690g/mL
Quantity 10kg
Color Undesignated
Melting Point 2.0°C to 4.0°C
Refractive Index 1.3672 to 1.3692
Water 0.1% max. (K.F.)
Infrared Spectrum Authentic
Boiling Point 90.0°C
Assay Percent Range 99%
Linear Formula (CH3O)2CO
Beilstein 03, 4
Fieser 02,149; 05,234; 12,201
Merck Index 15, 3263
Solubility Information Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies
Formula Weight 90.08
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Dimethyl carbonate
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger