Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dimethyl L-tartrate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
608-68-4
C6H10O6
178.14
MFCD00064437
PVRATXCXJDHJJN-QWWZWVQMSA-N
+-dimethyl l-tartrate,dimethyl l-tartrate,2r,3r-dimethyl 2,3-dihydroxysuccinate,dimethyl +-tartrate,l-+-tartaric acid dimethyl ester,dimethyl l-+-tartrate,1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate,butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester,dimethyl 2r,3r-2,3-dihydroxybutanedioate,l-tartaric acid, dimethyl ester
11851
dimethyl (2R,3R)-2,3-dihydroxybutanedioate
COC(=O)C(C(C(=O)OC)O)O
608-68-4
C6H10O6
178.14
MFCD00064437
PVRATXCXJDHJJN-QWWZWVQMSA-N
+-dimethyl l-tartrate,dimethyl l-tartrate,2r,3r-dimethyl 2,3-dihydroxysuccinate,dimethyl +-tartrate,l-+-tartaric acid dimethyl ester,dimethyl l-+-tartrate,1,4-dimethyl 2r,3r-2,3-dihydroxybutanedioate,butanedioic acid, 2,3-dihydroxy-2r,3r-, 1,4-dimethyl ester,dimethyl 2r,3r-2,3-dihydroxybutanedioate,l-tartaric acid, dimethyl ester
11851
dimethyl (2R,3R)-2,3-dihydroxybutanedioate
COC(=O)C(C(C(=O)OC)O)O
Melting Point 48°C to 50°C
Flash Point >112°C
Boiling Point 157°C to 159°C (15.0 hPa)
Specific Rotation + 21
Quantity 500g
Infrared Spectrum Authentic
Specific Rotation Condition + 21.00 (20.00°C c=10,H2O)
Packaging Glass bottle
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula [-CH(OH)CO2CH3]2
Beilstein 03, 510
Fieser 13,124
Solubility Information Solubility in water: soluble in water
Formula Weight 178.14
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material (+)-Dimethyl L-tartrate
Show More Show Less

EINECSNumber : 210-166-8