Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylethylhexadecylammonium bromide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
124-03-8
C20H44BrN
378.483
MFCD00011774
VUFOSBDICLTFMS-UHFFFAOYSA-M
Cetyldimethylethylammonium bromide
31280
ethyl-hexadecyl-dimethylazanium;bromide
CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]
124-03-8
C20H44BrN
378.483
MFCD00011774
VUFOSBDICLTFMS-UHFFFAOYSA-M
Cetyldimethylethylammonium bromide
31280
ethyl-hexadecyl-dimethylazanium;bromide
CCCCCCCCCCCCCCCC[N+](C)(C)CC.[Br-]
Phase transfer catalyst
Additional Information amine hydrobromide: < 0.5%
Amine < 0.2%
Assay Percent Range 99%
Linear Formula CH3(CH2)15N(CH3)2(C2H5)Br
Beilstein 04, III, 425
Infrared Spectrum Authentic
Merck Index 15, 2028
Packaging Glass bottle
Formula Weight 378.48
Physical Form Powder
Percent Purity ≥98.5%
Color White
pH 6 to 8 (1% soln.)
Quantity 100g
Chemical Name or Material Dimethylethylhexadecylammonium bromide, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 204-672-8

RTECSNumber : BQ5425000

TSCA : TSCA