Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylformamide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
68-12-2
C3H7NO
73.095
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide,dimethyl formamide,n,n-dimethylmethanamide,n-formyldimethylamine,formamide, n,n-dimethyl,dmf,dimethylformamid,dimetilformamide,dwumetyloformamid,dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.095
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide,dimethyl formamide,n,n-dimethylmethanamide,n-formyldimethylamine,formamide, n,n-dimethyl,dmf,dimethylformamid,dimetilformamide,dwumetyloformamid,dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 73.09
Grade BAKER ANALYZED™ ACS Reagent
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.95kg
Flash Point 58.00°C
Quantity 4L
Chemical Name or Material Dimethylformamide