Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylformamide, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum

Chemical Identifiers

68-12-2
C3H7NO
73.095
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

Specifically manufactured for use in organometallic and oligonucleotide synthesis

    For Use in Organic Synthesis
Formula Weight 73.09
Packaging Glass Bottle
Specific Gravity 1L = 0.95kg
Flash Point 58.00°C
Quantity 100mL
Chemical Name or Material Dimethylformamide
Grade BakerDRY™