Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylformamide, PHOTREX™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

68-12-2
C3H7NO
73.095
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
CHEBI:17741

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry
CAS 68-12-2
Molecular Formula C3H7NO
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES CN(C)C=O
IUPAC Name N,N-dimethylformamide
Molecular Weight (g/mol) 73.095
PubChem CID 6228
ChEBI CHEBI:17741
Formula Weight 73.09
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.95kg
Flash Point 58.00°C
Quantity 4L
Chemical Name or Material Dimethylformamide
Grade PHOTREX™ Reagent
Show More Show Less