Alfa Aesar™ Dimethylgermanium dichloride, 98+%

Manufacturer: Alfa Aesar™ 3410702

Quantity 0.5g

 View more versions of this product

Catalog No. AA3410702

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1529-48-2
    Formula Weight 173.57
    MDL Number MFCD00013588
    UN Number UN2924
    Synonym Dichlorodimethylgermane
    Chemical Name or Material Dimethylgermanium dichloride

    Specifications

    Formula Weight 173.57
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Boiling Point 124°C
    Melting Point −22°C
    Synonym Dichlorodimethylgermane
    Flash Point 32° (90°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Sensitivity Moisture Sensitive
    Physical Form Liquid
    UN Number UN2924
    Density 1.51g/mL
    Refractive Index 1.460
    MDL Number MFCD00013588
    Quantity 0.5g
    Health Hazard 1 Danger
    CAS 1529-48-2
    Chemical Name or Material Dimethylgermanium dichloride
    Assay ≥98%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit