Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylglyoxime, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
95-45-4
C4H8N2O2
116.12
MFCD00002117
OFZZNQXYRRSXOI-ONEGZZNKSA-N
biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky
5356010
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
CC(=C(C)N=O)NO
95-45-4
C4H8N2O2
116.12
MFCD00002117
OFZZNQXYRRSXOI-ONEGZZNKSA-N
biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky
5356010
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
CC(=C(C)N=O)NO

Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger

Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger

Insoluble Matter 0.05% max. (in alcohol)
Melting Point 239.0°C to 242.0°C
Sublimation Point >70 °C
Infrared Spectrum Authentic
Sulfated Ash 0.1% max.
Packaging Plastic bottle
Quantity 500g
Color White
Assay Percent Range 99+%
Linear Formula CH3C(=NOH)C(=NOH)CH3
Beilstein 01,772
Merck Index 15,327
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Formula Weight 116.12
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material Dimethylglyoxime, 99+%
Show More Show Less

GHS H Statement
Flammable solid.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediate

GHS Signal Word: Danger

EINECSNumber : 202-420-1

RTECSNumber : EK2975000

TSCA : TSCA