Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethylglyoxime, ACS

Manufacturer:  Alfa Aesar™ 3331030

Quantity 250g

 View more versions of this product

Catalog No. AA3331030


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

95-45-4
MFCD00002117
14,3246
Dimethylglyoxime
116.12
506731
2,3-Butanedione dioxime; Diacetyl dioxime
ACS Grade

Specifications

White to off-white
250g
Powder
MFCD00002117
14,3246
Soluble in alcohol, acetone, ether
ACS Grade
≈240°C (dec.)
95-45-4
116.12
506731
2,3-Butanedione dioxime; Diacetyl dioxime
Dimethylglyoxime

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EK2975000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit