Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylglyoxime, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
95-45-4
C4H8N2O2
116.12
OFZZNQXYRRSXOI-ONEGZZNKSA-N
biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky
5356010
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
CC(=C(C)N=O)NO
95-45-4
C4H8N2O2
116.12
OFZZNQXYRRSXOI-ONEGZZNKSA-N
biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky
5356010
N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
CC(=C(C)N=O)NO

High quality chemicals for laboratory and specialized industrial use

Packaging Poly Bottle
Specific Gravity 1L = 1.37kg
Quantity 500g
Formula Weight 116.12
Physical Form Powder
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Dimethylglyoxime