Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylglyoxime Sodium Salt, Spectrum™

Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Fiber Drum
Poly Bottle
75006-64-3
75006-64-3
Packaging Fiber Drum
Insoluble Matter 0.0005
Quantity 25 kg
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.