Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimethyloxaloylglycine, 98%

Click to view available options
Quantity:
10mg
50mg
100mg
89464-63-1
C6H9NO5
175.14
MFCD05865098
BNJOZDZCRHCODO-UHFFFAOYSA-N
dmog,dimethyloxalylglycine,dimethyloxaloylglycine,dimethyloxallyl glycine,methyl n-methoxy oxo acetyl glycinate,methyl 2-2-methoxy-2-oxoacetamido acetate,methyl 2-2-methoxy-2-oxoethyl amino-2-oxoacetate,glycine, n-methoxyoxoacetyl-, methyl ester,n-2-methoxy-2-oxoacetyl glycine methyl ester,glycine,n-2-methoxy-2-oxoacetyl-, methyl ester
560326
methyl 2-[(2-methoxy-2-oxoethyl)amino]-2-oxoacetate
COC(=O)CNC(=O)C(=O)OC
89464-63-1
C6H9NO5
175.14
MFCD05865098
BNJOZDZCRHCODO-UHFFFAOYSA-N
dmog,dimethyloxalylglycine,dimethyloxaloylglycine,dimethyloxallyl glycine,methyl n-methoxy oxo acetyl glycinate,methyl 2-2-methoxy-2-oxoacetamido acetate,methyl 2-2-methoxy-2-oxoethyl amino-2-oxoacetate,glycine, n-methoxyoxoacetyl-, methyl ester,n-2-methoxy-2-oxoacetyl glycine methyl ester,glycine,n-2-methoxy-2-oxoacetyl-, methyl ester
560326
methyl 2-[(2-methoxy-2-oxoethyl)amino]-2-oxoacetate
COC(=O)CNC(=O)C(=O)OC
Formula Weight 175.14
Physical Form Solid
Percent Purity 98%
Color White
Quantity 100mg
Chemical Name or Material Dimethyloxaloylglycine

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C