Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylphosphoramidous Dichloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
683-85-2
C2H6Cl2NP
145.96
MFCD00013613
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
dimethylphosphoramidous dichloride,dichloro dimethylamino phosphine,phosphoramidous dichloride, dimethyl,dimethylamino dichlorophosphine,dimethylphosphoramidousdichloride,me2npcl2,dimethylaminodichlorphosphine,dimethylaminodichlorophosphine,ch3 2npcl2,dichlorophosphanyl dimethylamine
136483
N-dichlorophosphanyl-N-methylmethanamine
CN(C)P(Cl)Cl
683-85-2
C2H6Cl2NP
145.96
MFCD00013613
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
dimethylphosphoramidous dichloride,dichloro dimethylamino phosphine,phosphoramidous dichloride, dimethyl,dimethylamino dichlorophosphine,dimethylphosphoramidousdichloride,me2npcl2,dimethylaminodichlorphosphine,dimethylaminodichlorophosphine,ch3 2npcl2,dichlorophosphanyl dimethylamine
136483
N-dichlorophosphanyl-N-methylmethanamine
CN(C)P(Cl)Cl
Specific Gravity 1.271
Flash Point −23°C
Quantity 25mL
Boiling Point 150.0°C
Color Colorless to Yellow
Density 1.2710g/mL
Refractive Index 1.5040 to 1.5060 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
NMR 96.0% min.
Assay Percent Range 97%
Linear Formula (CH3)2NPCl2
Solubility Information Solubility in water: reacts
Formula Weight 145.96
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dimethylphosphoramidous dichloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger