Alfa Aesar™ Dimethylsuberimidate dihydrochloride

Crosslinking of proteins and poly(allylamine)

Manufacturer: Alfa Aesar™ 4324409

Quantity 10g

 View more versions of this product

Catalog No. AA4324409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34490-86-3
    Molecular Formula C10H20N2O2·2HCl
    Formula Weight 273.2
    MDL Number MFCD00012574
    Chemical Name or Material Dimethylsuberimidate dihydrochloride

    Specifications

    Formula Weight 273.2
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H20N2O2·2HCl
    MDL Number MFCD00012574
    Sensitivity Moisture Sensitive
    Health Hazard 1 Warning
    TSCA TSCA
    Melting Point 213° to 214°C
    CAS 34490-86-3
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Crystalline
    Quantity 10g
    Solubility Information Soluble in water
    Chemical Name or Material Dimethylsuberimidate dihydrochloride

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit