Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyltin dichloride, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
753-73-1
C2H6Cl2Sn
219.66
MFCD00000517
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
dimethyltin dichloride,dimethyldichlorotin,stannane, dichlorodimethyl,dichlorodimethyltin,dimethyltindichloride,dimethyldichlorostannane,dichloro dimethyl stannane,tin, dimethyl-, dichloride,cotin 210,dichlorid dimethylcinicity
12955
dichloro(dimethyl)stannane
C[Sn](C)(Cl)Cl
753-73-1
C2H6Cl2Sn
219.66
MFCD00000517
PKKGKUDPKRTKLJ-UHFFFAOYSA-L
dimethyltin dichloride,dimethyldichlorotin,stannane, dichlorodimethyl,dichlorodimethyltin,dimethyltindichloride,dimethyldichlorostannane,dichloro dimethyl stannane,tin, dimethyl-, dichloride,cotin 210,dichlorid dimethylcinicity
12955
dichloro(dimethyl)stannane
C[Sn](C)(Cl)Cl
Packaging Glass bottle
Color White to Yellow
Quantity 100g
Melting Point 103.0°C to 110.0°C
Infrared Spectrum Authentic
Boiling Point 188.0°C to 190.0°C
Assay Percent Range 94% min. (Argentometry)
Linear Formula (CH3)2Sn(Cl)2
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: 0.1 g/mL in methanol
Formula Weight 219.66
Physical Form Crystals
Percent Purity 95%
Chemical Name or Material Dimethyltin dichloride
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 212-039-2