Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylvinylchlorosilane, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1719-58-0
C4H9ClSi
120.65
MFCD00018090
XSDCTSITJJJDPY-UHFFFAOYSA-N
chlorodimethylvinylsilane,dimethylvinylchlorosilane,vinyldimethylchlorosilane,chloro dimethyl vinylsilane,silane, chloroethenyldimethyl,chlorodimethyl vinyl silane,unii-xja6n5221t,chloroethenyldimethyl-silane,chloro ethenyl dimethylsilane,chloro-dimethyl-ethenyl-silane
519368
chloro-ethenyl-dimethylsilane
C[Si](C)(C=C)Cl
1719-58-0
C4H9ClSi
120.65
MFCD00018090
XSDCTSITJJJDPY-UHFFFAOYSA-N
chlorodimethylvinylsilane,dimethylvinylchlorosilane,vinyldimethylchlorosilane,chloro dimethyl vinylsilane,silane, chloroethenyldimethyl,chlorodimethyl vinyl silane,unii-xja6n5221t,chloroethenyldimethyl-silane,chloro ethenyl dimethylsilane,chloro-dimethyl-ethenyl-silane
519368
chloro-ethenyl-dimethylsilane
C[Si](C)(C=C)Cl
Specific Gravity 0.88
Refractive Index 1.4140 to 1.4170
Packaging Glass bottle
Boiling Point 82.0°C
Color Amber to Beige
Quantity 5g
Infrared Spectrum Authentic
Flash Point −4°C
Density 0.8800g/mL
Assay Percent Range 96% min. (GC)
Linear Formula Cl(CH3)2SiCHCH2
Solubility Information Solubility in water: reacts with water
Formula Weight 120.65
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dimethylvinylchlorosilane, 97%
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 217-007-1

TSCA : TSCA