Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylzinc, 1.2M solution in toluene, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
544-97-8,108-88-3
C2H6Zn
95.45
MFCD00014854
JRPGMCRJPQJYPE-UHFFFAOYSA-N
dimethylzinc,zinc, dimethyl,dimethyl zinc,methylzinc,unii-8h6r0n8w2f,ch3 2zn,znme2,zinc methyl,dimethylzink,hsdb 709
11010
zinc;carbanide
[CH3-].[CH3-].[Zn+2]
544-97-8,108-88-3
C2H6Zn
95.45
MFCD00014854
JRPGMCRJPQJYPE-UHFFFAOYSA-N
dimethylzinc,zinc, dimethyl,dimethyl zinc,methylzinc,unii-8h6r0n8w2f,ch3 2zn,znme2,zinc methyl,dimethylzink,hsdb 709
11010
zinc;carbanide
[CH3-].[CH3-].[Zn+2]
Appearance of Solution Clear colorless
Linear Formula Zn(CH3)2
Concentration 1.2M (based on Zn) exact strength on the certificate of analysis
Density 0.9300g/mL
Merck Index 15, 3289
Formula Weight 95.45
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction. Other solubilities: soluble in hydrocarbons
Specific Gravity 0.93
Quantity 100mL
Chemical Name or Material Dimethylzinc, 1.2M Solution in Toluene
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of damaging the unborn child. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.

Danger