Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylzinc, 96%

Catalog No. aa3956314
Click to view available options
Quantity:
25 g
Packaging:
Cylinder

CAS: 544-97-8 | C2H6Zn | 95.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-97-8
Molecular Formula C2H6Zn
Molecular Weight (g/mol) 95.45
MDL Number MFCD00014854
InChI Key JRPGMCRJPQJYPE-UHFFFAOYSA-N
Synonym dimethylzinc, zinc, dimethyl, dimethyl zinc, methylzinc, unii-8h6r0n8w2f, ch3 2zn, znme2, zinc methyl, dimethylzink, hsdb 709
PubChem CID 11010
IUPAC Name zinc;carbanide
SMILES [CH3-].[CH3-].[Zn+2]

Specifications

Melting Point -40°C
Density 1.386 g/mL at 20°C
Boiling Point 46°C
Flash Point -18°C (-0.4°F)
Packaging Cylinder
Quantity 25 g
UN Number UN3394
Beilstein 3587195
Sensitivity Air and moisture sensitive
Merck Index 14,3263
Formula Weight 95.44
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Dimethylzinc
Show More Show Less
Hazard Category H250-H260-H314-H335
Hazard Statement GHS H Statement
H250-H260-H252-H314-H318
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P222-P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 208-884-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.