Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Dimidium bromide, 95%

Click to view available options
Quantity:
0.25g
1g
518-67-2
C20H18BrN3
380.289
MFCD00011757
MQOKYEROIFEEBH-UHFFFAOYSA-N
dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553
68207
5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
C[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[Br-]
518-67-2
C20H18BrN3
380.289
MFCD00011757
MQOKYEROIFEEBH-UHFFFAOYSA-N
dimidium bromide,trypadine,3,8-diamino-5-methyl-6-phenylphenanthridinium bromide,phenanthridinium 1553,phenanthridinium compound 1553,phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide,2,7-diamino-10-methyl-9-phenylphenanthridinium bromide,3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide,phenanthridium 1553
68207
5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
C[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[Br-]
Applications
Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Melting Point 230°C (decomposition)
Quantity 0.25g
Formula Weight 380.31
Percent Purity 95%
Chemical Name or Material Dimidium bromide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SF7960500