Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimidium bromide, 95%

Catalog No. AAB2481803
Click to view available options
Quantity:
0.25 g
1 g

CAS: 518-67-2 | C20H18BrN3 | 380.29 g/mol

Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimidium bromide acts as an inhibitor of L-cell growth. It is also used as a fluorescent probe for nucleic acids. It is also useful in the determination of cationic surfactants. It is also used as an intercalating agent.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-67-2
Molecular Formula C20H18BrN3
Molecular Weight (g/mol) 380.29
MDL Number MFCD00011757
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
PubChem CID 68207
IUPAC Name 5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point 230°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 380.31
Percent Purity 95%
Chemical Name or Material Dimidium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-256-7
RTECSNumber SF7960500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.