Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIMT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113524
Description
DIMT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3'-end of 18S rRNA in the 40S particle.
Specifications
DIMT1 | |
Polyclonal | |
Unconjugated | |
DIMT1 | |
1500031M22Rik; 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; 18S rRNA dimethylase; AV111347; DIM1; DIM1 dimethyladenosine transferase 1 homolog; DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae); DIM1 dimethyladenosine transferase 1-like; DIM1 dimethyladenosine transferase 1-like (S. cerevisiae); dimethyladenosine transferase; Dimt1; Dimt1l; HSA9761; HUSSY-05; HUSSY5; probable 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase; Probable 18S rRNA dimethylase; probable dimethyladenosine transferase; Probable S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase; RGD1311752; S-adenosylmethionine-6-N',N'-adenosyl(rRNA) dimethyltransferase | |
Rabbit | |
Protein G | |
RUO | |
27292, 66254 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9D0D4, Q9UNQ2 | |
DIMT1 | |
Recombinant Human Probable dimethyladenosine transferase protein (1-313AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction