Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dinitrodurene, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
5465-13-4
C10H12N2O4
224.216
AEPQXGFMAZTUEA-UHFFFAOYSA-N
1,2,4,5-tetramethyl-3,6-dinitro-benzene,dinitrotrodurene,3,2,4,5-tetramethylbenzene,2,3,5,6-tetramethyl-p-dinitrobenzene,1,4,5-tetramethyl-3,6-dinitrobenzene,benzene,2,4,5-tetramethyl-3,6-dinitro,1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
79593
1,2,4,5-tetramethyl-3,6-dinitrobenzene
CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C
5465-13-4
C10H12N2O4
224.216
AEPQXGFMAZTUEA-UHFFFAOYSA-N
1,2,4,5-tetramethyl-3,6-dinitro-benzene,dinitrotrodurene,3,2,4,5-tetramethylbenzene,2,3,5,6-tetramethyl-p-dinitrobenzene,1,4,5-tetramethyl-3,6-dinitrobenzene,benzene,2,4,5-tetramethyl-3,6-dinitro,1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
79593
1,2,4,5-tetramethyl-3,6-dinitrobenzene
CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C
Quantity 25g
Packaging Glass bottle
Melting Point 208.0°C to 213.0°C
Infrared Spectrum Authentic
Linear Formula (CH3)4C6(NO2)2
Formula Weight 224.22
Percent Purity 97%
Chemical Name or Material Dinitrodurene

GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Store in a well-ventilated place. Keep container tightly closed.

Danger