Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioctyl phthalate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
25kg
117-81-7
C24H38O4
390.564
MFCD00009493
BJQHLKABXJIVAM-UHFFFAOYSA-N
dehp,bis 2-ethylhexyl phthalate,di 2-ethylhexyl phthalate,di 2-ethylhexyl phthalate,bis 2-ethylhexyl phthalate,diethylhexyl phthalate,di-sec-octyl phthalate,octyl phthalate,fleximel,octoil
8343
CHEBI:17747
bis(2-ethylhexyl) benzene-1,2-dicarboxylate
CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC
117-81-7
C24H38O4
390.564
MFCD00009493
BJQHLKABXJIVAM-UHFFFAOYSA-N
dehp,bis 2-ethylhexyl phthalate,di 2-ethylhexyl phthalate,di 2-ethylhexyl phthalate,bis 2-ethylhexyl phthalate,diethylhexyl phthalate,di-sec-octyl phthalate,octyl phthalate,fleximel,octoil
8343
CHEBI:17747
bis(2-ethylhexyl) benzene-1,2-dicarboxylate
CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC
Vapor Pressure 1.8mbar at 200°C
Color Undesignated
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Water 0.1% max.
Density 0.9810g/mL
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Beilstein 09, IV, 3180
Merck Index 15, 2868
Formula Weight 390.55
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Dioctyl phthalate
Show More Show Less

GHS H Statement:
May damage fertility. May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger