Learn More
Thermo Scientific™ Dionex™ ASE™ 150/350 Dionium Extraction Cell Bodies and End Caps
This is the replacement for the cylindrical body of the Dionium extraction cell in your Thermo Scientific™ Dionex ASE 150 or 350 Accelerated Solvent Extractor.
$563.01
Specifications
For Use With (Equipment) ASE 150 and 350
Material Dionium
Type Extraction Cell Body
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
50-134-5478 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific™
068265
Extraction Cell Body; 66 mL Each for $563.01
Add to cart
Description
Description
Need to replace the cylindrical body of the Dionium extraction cell in your Thermo Scientific™ Dionex™ ASE™ 150 or 350 Accelerated Solvent Extractor? Available in 66mL and 100mL, these Dionex ASE 150/350 Dionium Extraction Cell Bodies are resistant to low concentrations of mineral acids and strong bases. This part does not include extraction cell caps or O-rings.
Dionex ASE systems reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices. These systems feature extraction cells with finger-tight cell caps that provide a compression seal for high-pressure closure that's easy to use.
Specifications
Specifications
ASE 150 and 350
Dionium
Extraction Cell Body
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.