Learn More
Thermo Scientific™ IPTG Promotion
Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.
Manufacturer:  Thermo Scientific™ R0392
 View more versions of this product
Catalog No. FERR0392PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.
It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.
Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22µm membrane-filtered, formulated for direct use in blue/white colony screening.
Specifications
Specifications
direct use in blue/white colony screening.
IPTG, dioxane-free
5g