Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dip Slides

Make colony counting simple for aerobic bacteria, yeast and fungi with the easily disposable Thermo Scientific™ Dip-Slides™.

$54.18 - $71.19

Specifications

Format Dip Slide
Manufacturing Origin United States
Storage Requirements Store at 15-25°C
View More Specs

Products 4
Catalog Number Mfr. No. Description Format Quantity Manufacturing Origin Price Quantity & Availability  
Catalog Number Mfr. No. Description Format Quantity Manufacturing Origin Price Quantity & Availability  
R65205
View Documents
Thermo Scientific™
R65205
Dip Slide Modified CLED Medium / EMB Agar Dip Slide 10/Pk. United States Pack of 10 for $56.18
Add to cart
 
OXDS119
View Documents
Thermo Scientific™
R65119
Dip Slide MacConkey Agar No.3 / CLED Medium Dip Slide 10/Pk. United States Pack of 10 for $54.18
Add to cart
 
OXDS154
View Documents
Thermo Scientific™
R65154
Dip Slide Nutrient Agar w/TTC Agar / Malt Extract Agar Dip Slide 10/Pk. United States Pack of 10 for $71.19
Add to cart
 
OXDS146
View Documents
Thermo Scientific™
R65146
Dip Slide Nutrient Agar w/TTC Agar Dip Slide 10/Pk. United States Pack of 10 for $66.57
Add to cart
 
Description

Description

Make colony counting simple for aerobic bacteria, yeast and fungi with the easily disposable Thermo Scientific™ Dip-Slides™. Simple-to-use for the operator and where laboratory facilities are limited, the Dip Slide is coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective.

Originally designed to determine bacterial counts on urine specimens, dip slides have been modified over the years by making the device double-sided, with one side a medium for Gram positive species and the other side a medium for Gram negatives. With these dip slides, the process for colony counting is time- and cost-effective, allowing exposure of the specimens to two different media simultaneously.
  • Slide is made of easily disposable plastic with raised edges to ensure an even thickness of culture medium
  • Molded grid allows for easier colony counting
  • Slide is shaped to ensure drainage of fluid from the slide
  • Cap forms a convenient handle by which it may be held without risk of touching the culture medium
  • Bacteria, fungal and yeast charts are included, making examining growth simple for the operator and where laboratory facilities are limited
Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications

Specifications

Dip Slide
United States
Store at 15-25°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit