Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIP13B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530669
Description
Recommended positive controls: NT2D1, IMR32. Predicted reactivity: Mouse (94%), Rat (93%), Xenopus laevis (87%), Pig (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Required for the regulation of cell proliferation in response to extracellular signals mediated by an early endosomal compartment. Links Rab5 to nuclear signal transduction.
Specifications
DIP13B | |
Polyclonal | |
Unconjugated | |
APPL2 | |
adapter protein containing PH domain, PTB domain and leucine zipper motif 2; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2; Appl2; DCC-interacting protein 13-beta; DIP13 beta; DIP13B; dip13-beta; Dip3 beta; Dip3b; RGD1563028 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55198 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.6 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NEU8 | |
APPL2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 225 of Human DIP13B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction