Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentaerythritol 90%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
Packaging:
Plastic bottle
126-58-9
C10H22O7
254.28
MFCD00004691
TXBCBTDQIULDIA-UHFFFAOYSA-N
dipentaerythritol,dipentek,2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol,bis pentaerythritol,1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl,2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol,1,3-propanediol, 2,2'-oxybis-methylene,2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol
31352
2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)COCC(CO)(CO)CO)O
126-58-9
C10H22O7
254.28
MFCD00004691
TXBCBTDQIULDIA-UHFFFAOYSA-N
dipentaerythritol,dipentek,2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol,bis pentaerythritol,1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl,2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol,1,3-propanediol, 2,2'-oxybis-methylene,2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol
31352
2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)COCC(CO)(CO)CO)O
Boiling Point 356°C
Flash Point >150°C
Melting Point 215°C to 225°C
Color White
Quantity 500g
Packaging Plastic bottle
Infrared Spectrum Authentic
Assay Percent Range 85+%
Linear Formula (HOCH2)3CCH2OCH2C(CH2OH)3
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: 0.99g/100 mL water (50°C),2.1g/L methanol (25°C),0.6g/L ethanol (25°C)
Formula Weight 254.28
Physical Form Crystalline Powder
Percent Purity ≥88%
Grade Technical
Chemical Name or Material Dipentaerythritol, 90%
Show More Show Less

HYGROSCOPIC

EINECSNumber : 204-794-1

TSCA : TSCA