Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentene, pure, fraction of terpene hydrocarbons, Acros Organics

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
68956-56-9
C10H16
136.23
MFCD00062992
spermine,gerontine,musculamine,neuridine,spermin,4,9-diaza-1,12-dodecanediamine,diaminopropyltetramethylenediamine,n,n'-bis 3-aminopropyl-1,4-butanediamine,spermine, puriss,1,4-butanediamine, n,n'-bis 3-aminopropyl
68956-56-9
C10H16
136.23
MFCD00062992
spermine,gerontine,musculamine,neuridine,spermin,4,9-diaza-1,12-dodecanediamine,diaminopropyltetramethylenediamine,n,n'-bis 3-aminopropyl-1,4-butanediamine,spermine, puriss,1,4-butanediamine, n,n'-bis 3-aminopropyl
Packaging Glass bottle
Quantity 2.5L
Boiling Point 174.0°C
Color Undesignated
Refractive Index 1.4730 to 1.4780 (20°C, 589nm)
Flash Point 42°C
Density 0.8680g/mL
Specific Gravity 0.868
Formula Weight 136.23
Physical Form Liquid
Grade Pure
Chemical Name or Material Dipentene
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Harmful if swallowed.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 273-309-3