Alfa Aesar™ Dipentene, tech.

Manufacturer: Alfa Aesar™ 45805AD

Quantity 50mL

 View more versions of this product

Catalog No. AA45805AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 138-86-3
    Molecular Formula C10H16
    Linear Formula CH3C6H8C(CH3)=CH2
    Formula Weight 136.24
    MDL Number MFCD00062992
    UN Number UN2052
    Beilstein 3587825
    Merck Index 14,5493
    Synonym (+/-)-Limonene; (+/-)-p-Mentha-1,8-diene
    Chemical Name or Material Dipentene
    Grade Technical Grade

    Specifications

    Formula Weight 136.24
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H16
    Merck Index 14,5493
    Physical Form Liquid
    Linear Formula CH3C6H8C(CH3)=CH2
    Flash Point 43°C (109°F)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIPENTENE
    RTECS Number OS8100000
    TSCA TSCA
    Refractive Index 1.474
    CAS 138-86-3
    UN Number UN2052
    Synonym (+/-)-Limonene; (+/-)-p-Mentha-1,8-diene
    MDL Number MFCD00062992
    Boiling Point 170° to 180°C
    Quantity 50mL
    Health Hazard 2 GHS H Statement
    H226-H315-H317
    Flammable liquid and vapor.
    Causes skin irritation.
    May cause an allergic skin reaction.
    Density 0.85g/mL
    Beilstein 3587825
    Health Hazard 1 Warning
    Chemical Name or Material Dipentene
    Grade Technical Grade

    GHS H Statement
    H226-H315-H317
    Flammable liquid and vapor.
    Causes skin irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit