Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipentyl phthalate, 97%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
131-18-0
C18H26O4
306.4
IPKKHRVROFYTEK-UHFFFAOYSA-N
dipentyl phthalate,di-n-pentyl phthalate,diamyl phthalate,amyl phthalate,amoil,di-n-amyl phthalate,di-n-pentylphthalate,phthalic acid, dipentyl ester,phthalic acid diamyl ester,1,2-benzenedicarboxylic acid, dipentyl ester
8561
CHEBI:34680
dipentyl benzene-1,2-dicarboxylate
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
131-18-0
C18H26O4
306.4
IPKKHRVROFYTEK-UHFFFAOYSA-N
dipentyl phthalate,di-n-pentyl phthalate,diamyl phthalate,amyl phthalate,amoil,di-n-amyl phthalate,di-n-pentylphthalate,phthalic acid, dipentyl ester,phthalic acid diamyl ester,1,2-benzenedicarboxylic acid, dipentyl ester
8561
CHEBI:34680
dipentyl benzene-1,2-dicarboxylate
CCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCC
Boiling Point 342.0°C
Refractive Index 1.4885 to 1.4905
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Specific Gravity 1.023
Melting Point -55.0°C
Density 1.0230g/mL
Quantity 25g
Assay Percent Range 96% min. (GC)
Linear Formula (C(O)OC5H11)2C6H4
Formula Weight 306.4
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Dipentyl phthalate
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 205-017-9