Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenhydramine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
147-24-0
C17H22ClNO
291.82
PCHPORCSPXIHLZ-UHFFFAOYSA-N
hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
147-24-0
C17H22ClNO
291.82
PCHPORCSPXIHLZ-UHFFFAOYSA-N
hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1

DI160, 147-24-0, C17H21NO•HCl

Spectrum™ Diphenhydramine Hydrochloride, USP is an antihistamine that includes sedative attributes to treat allergies. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 50 kg
CAS Max % 1
Percent Purity 98 to 102%