Alfa Aesar™ Diphenic anhydride, 98%

Manufacturer: Alfa Aesar™ B2341509

Quantity 10g

 View more versions of this product

Catalog No. AAB2341509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6050-13-1
    Molecular Formula C14H8O3
    Formula Weight 224.22
    MDL Number MFCD00004993
    Beilstein 171255
    Synonym 2,2'-Biphenyldicarboxylic anhydride
    Chemical Name or Material Diphenic anhydride

    Specifications

    Formula Weight 224.22
    MDL Number MFCD00004993
    Beilstein 171255
    Melting Point 223° to 228°C
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C14H8O3
    CAS 6050-13-1
    Synonym 2,2'-Biphenyldicarboxylic anhydride
    Health Hazard 1 Warning
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Diphenic anhydride
    Assay 98%

    Description

    Applications
    Diphenic anhydride when treated with quinaldine and zinc chloride it produces quinodiphenone. It is used in the determination of fatty alcohol ethoxylates as derivatization reagent. It is used as medicine, dye and plating solution intermediates.

    Solubility
    Decomposes in water.

    Notes
    Moisture Sensitive. Store under dry inert gas. Store away from moisture and oxidizing agents. Protect from humidity and water.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit