Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Diphenyl-γ-butyrolactone, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
956-89-8
C16H14O2
238.286
MFCD00005393
IRQJWIUKHLVISG-UHFFFAOYSA-N
3,3-diphenyldihydrofuran-2 3h-one,alpha,alpha-diphenyl-gamma-butyrolactone,dihydro-3,3-diphenyl-2 3h-furanone,3,3-diphenyl-3,4,5-trihydrofuran-2-one,3,3-diphenyltetrahydrofuran-2-one,2 3h-furanone, dihydro-3,3-diphenyl,.alpha.,.alpha.-diphenyl-.gamma.-butyrolactone,4-hydroxy-2,2-diphenylbutyric acid,a-lactone
70393
3,3-diphenyloxolan-2-one
C1COC(=O)C1(C2=CC=CC=C2)C3=CC=CC=C3
956-89-8
C16H14O2
238.286
MFCD00005393
IRQJWIUKHLVISG-UHFFFAOYSA-N
3,3-diphenyldihydrofuran-2 3h-one,alpha,alpha-diphenyl-gamma-butyrolactone,dihydro-3,3-diphenyl-2 3h-furanone,3,3-diphenyl-3,4,5-trihydrofuran-2-one,3,3-diphenyltetrahydrofuran-2-one,2 3h-furanone, dihydro-3,3-diphenyl,.alpha.,.alpha.-diphenyl-.gamma.-butyrolactone,4-hydroxy-2,2-diphenylbutyric acid,a-lactone
70393
3,3-diphenyloxolan-2-one
C1COC(=O)C1(C2=CC=CC=C2)C3=CC=CC=C3
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Color White to Brown
Melting Point 75°C to 79°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 238.29
Physical Form Crystalline Powder
Percent Purity ≥94%
Grade Technical
Chemical Name or Material α, α-Diphenyl-γ-butyrolactone, tech.
Show More Show Less

EINECSNumber : 213-499-7

TSCA : TSCA