Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl chlorophosphate, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
2524-64-3
C12H10ClO3P
268.63
MFCD00003030
BHIIGRBMZRSDRI-UHFFFAOYSA-N
diphenyl chlorophosphate,diphenyl phosphorochloridate,diphenylchlorophosphate,phosphorochloridic acid, diphenyl ester,chlorodiphenyl phosphate,o,o-diphenyl chlorophosphate,chlorodiphenoxyphosphine oxide,diphenoxychlorophosphine oxide,diphenyl chlorophosphonate,diphenylphosphoric acid monochloride
75654
[chloro(phenoxy)phosphoryl]oxybenzene
C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl
2524-64-3
C12H10ClO3P
268.63
MFCD00003030
BHIIGRBMZRSDRI-UHFFFAOYSA-N
diphenyl chlorophosphate,diphenyl phosphorochloridate,diphenylchlorophosphate,phosphorochloridic acid, diphenyl ester,chlorodiphenyl phosphate,o,o-diphenyl chlorophosphate,chlorodiphenoxyphosphine oxide,diphenoxychlorophosphine oxide,diphenyl chlorophosphonate,diphenylphosphoric acid monochloride
75654
[chloro(phenoxy)phosphoryl]oxybenzene
C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl
Density 1.2900g/mL
Refractive Index 1.5485 to 1.5505
Boiling Point 314°C to 316°C (272.0mmHg)
Flash Point >112°C
Specific Gravity 1.29
Viscosity 14 mPa.s (25°C)
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25mL
Assay Percent Range 98%
Linear Formula (C6H5O)2P(O)Cl
Beilstein 03, 179
Fieser 01,345; 02,180; 03,133; 11,223
Merck Index 15, 3367
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents
Formula Weight 268.63
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Diphenyl chlorophosphate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger