Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diphenyl cyanocarbonimidate, 97%

Click to view available options
Quantity:
1g
5g
25g
79463-77-7
C14H10N2O2
238.246
MFCD00010380
SLIKWVTWIGHFJE-UHFFFAOYSA-N
diphenyl n-cyanocarbonimidate,diphenyl cyanocarbonimidate,diphenoxymethylenecyanamide,n-cyano-diphenyl imidocarbonate,diphenylcyanocarbonimidate,cyanocarbonimidia acid diphenyl aester,n-cyanocarbonimidic acid diphenyl ester,bis phenoxy methylidenecyanamide,cyano diphenoxymethylidene amine,3,3-diphenoxy-2-azaprop-2-enenitrile
688090
diphenoxymethylidenecyanamide
C1=CC=C(C=C1)OC(=NC#N)OC2=CC=CC=C2
79463-77-7
C14H10N2O2
238.246
MFCD00010380
SLIKWVTWIGHFJE-UHFFFAOYSA-N
diphenyl n-cyanocarbonimidate,diphenyl cyanocarbonimidate,diphenoxymethylenecyanamide,n-cyano-diphenyl imidocarbonate,diphenylcyanocarbonimidate,cyanocarbonimidia acid diphenyl aester,n-cyanocarbonimidic acid diphenyl ester,bis phenoxy methylidenecyanamide,cyano diphenoxymethylidene amine,3,3-diphenoxy-2-azaprop-2-enenitrile
688090
diphenoxymethylidenecyanamide
C1=CC=C(C=C1)OC(=NC#N)OC2=CC=CC=C2
Applications
Diphenyl N-cyanocarbonimidate has been used in the preparation of N-substituted 2-amino-4H-3,1-benzoxazines, cyanoguanidine derivatives, oxadiazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep under a nitrogen blanket. Store away from strong oxidizing agents, strong acids, strong bases, moist air, water.
Sensitivity Moisture sensitive
Quantity 1g
Melting Point 155°C to 158°C
Linear Formula (C6H5O)2C=NCN
Solubility Information Insoluble in water.
Formula Weight 238.25
Percent Purity 97%
Chemical Name or Material Diphenyl cyanocarbonimidate

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

EINECSNumber : 427-300-8

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen