Alfa Aesar™ Diphenyl selenide

Manufacturer: Alfa Aesar™ 6811803

Quantity 1g

 View more versions of this product

Catalog No. AA6811803

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Chemical Identifiers

    CAS 1132-39-4
    Molecular Formula (C6H5)2Se
    Formula Weight 233.17
    MDL Number MFCD00014067
    UN Number UN3440
    Synonym Phenyl selenide
    Chemical Name or Material Diphenyl selenide

    Specifications

    Formula Weight 233.17
    Health Hazard 3 GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Odor Unpleasant
    Molecular Formula (C6H5)2Se
    Density 1.338g/mL
    Flash Point >110°C (230°F)
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00014067
    Boiling Point 165° to 167°C (12mmHg)
    Melting Point 3°C
    Quantity 1g
    UN Number UN3440
    Synonym Phenyl selenide
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.
    CAS 1132-39-4
    Health Hazard 1 Danger
    Chemical Name or Material Diphenyl selenide

    GHS H Statement
    H301-H331-H373
    Toxic if swallowed.
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P261-P301+P310-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit